Assay Details
| Gene Symbol: Gjc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_221997.7 | XP_221997.5 | 1 - 2 | 1572 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_221997.7 transcripts.
Set Membership: |
|

Gjc3
Genomic Map