Assay Details
| Gene Symbol: Mrps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025408.1 | NP_001020579.1 | 1 - 2 | 174 | 103 | |
| GenBank mRNA | BC097381.1 | - | 1 - 2 | 174 | 7445826 | 103 |
| FQ210488.1 | - | 1 - 2 | 157 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps25
Genomic Map