Assay Details
| Gene Symbol: Lbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109244.1 | NP_001102714.1 | 1 - 2 | 261 | 119 | |
| Gene Symbol: LOC103692168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763083.1 | XP_008761305.1 | 1369 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lbx2
Genomic Map