Assay Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107214.1 | NP_001100684.1 | 4 - 5 | 1107 | 57 | |
| XM_006250898.2 | XP_006250960.1 | 4 - 5 | 1179 | 57 | ||
| XM_006250899.2 | XP_006250961.1 | 4 - 5 | 1178 | 57 | ||
| GenBank mRNA | BC158815.1 | - | 2 - 3 | 180 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slain2
Genomic Map