Assay Details
| Gene Symbol: LOC100911069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290110.1 | NP_001277039.1 | 3 - 4 | 317 | 67 | |
| XM_006227672.2 | XP_006227734.1 | 2 - 3 | 385 | 67 | ||
| GenBank mRNA | BC159435.1 | - | 3 - 4 | 317 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911069
Genomic Map