Assay Details
| Gene Symbol: Gbf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191634.1 | NP_001178563.1 | 4 - 5 | 493 | 62 | |
| XM_006231496.2 | XP_006231558.1 | 4 - 5 | 567 | 62 | ||
| XM_006231497.2 | XP_006231559.1 | 4 - 5 | 567 | 62 | ||
| XM_006231498.2 | XP_006231560.1 | 4 - 5 | 567 | 62 | ||
| XM_006231500.2 | XP_006231562.1 | 4 - 5 | 567 | 62 | ||
| XM_006231501.2 | XP_006231563.1 | 4 - 5 | 567 | 62 | ||
| XM_006231502.2 | XP_006231564.1 | 4 - 5 | 567 | 62 | ||
| GenBank mRNA | FQ216888.1 | - | 4 - 5 | 546 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gbf1
Genomic Map