Assay Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013108.3 | NP_001013126.2 | 13 - 14 | 1851 | 110 | |
| GenBank mRNA | AW141789.1 | - | 3 - 4 | 438 | 110 | |
| BC083881.1 | - | 11 - 12 | 1736 | 7131418 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1040999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124792 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1040999 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1040999 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1040999 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1040999 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6652 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64617 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32532 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77474 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55294 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32929 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39720 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84714 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10917980 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917978 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917982 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917983 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10917969 | - | - |
| Rat Genome 230 2.0 Array | 1392985_at | - | - |
| Rat Genome 230 2.0 Array | 1375595_at | - | - |
| Rat Genome 230 2.0 Array | 1382306_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375973_at | - | - |
| Rat Genome 230 2.0 Array | 1375595_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374823_at | - | - |
| Rat Genome 230 2.0 Array | 1382306_at | - | - |
| Rat Genome 230 2.0 Array | 1375973_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374823_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392985_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392985_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374823_at | - | - |
| Affymetrix HT Rat Focus Array | 1392985_at | - | - |
| Affymetrix HT Rat Focus Array | 1374823_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375595_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375973_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375595_at | - | - |
| Affymetrix HT Rat Focus Array | 1382306_at | - | - |
| Affymetrix HT Rat Focus Array | 1382306_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375973_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800021730.rn.1 | TC0800002668.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800021725.rn.1 | TC0800002668.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800021728.rn.1 | TC0800002668.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800021727.rn.1 | TC0800002668.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800021729.rn.1 | TC0800002668.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374823_PM_at | - | - |
| Rat Expression Set 230 | 1375595_at | - | - |
| Rat Expression Set 230 | 1374823_at | - | - |
| Rat Expression Set 230 | 1375973_PM_at | - | - |
| Rat Expression Set 230 | 1375973_at | - | - |
| Rat Expression Set 230 | 1375595_PM_at | - | - |
| Rat Expression Set 230 | 1382306_at | - | - |
| Rat Expression Set 230 | 1392985_PM_at | - | - |
| Rat Expression Set 230 | 1392985_at | - | - |
| Rat Expression Set 230 | 1382306_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10917969 | - | - |
| Rat Genome U34 Set | rc_AA901067_at | - | - |
| Rat Genome U34 Set | rc_AI101226_at | - | - |
| Rat Genome U34 Set | rc_AI231433_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21156554 | - | - |
| ABI Rat Genome Survey Microarray | 21661361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361429 | - | - |

Arih1
Genomic Map