Assay Details
Gene Symbol: Fam96a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008327.1 | NP_001008328.1 | 2 - 3 | 419 | 69 | |
GenBank mRNA | BC086524.1 | - | 2 - 3 | 419 | 7303322 | 69 |
FQ210328.1 | - | 2 - 3 | 400 | 69 | ||
FQ215407.1 | - | 2 - 3 | 456 | 69 | ||
FQ216067.1 | - | 2 - 3 | 463 | 69 | ||
FQ216458.1 | - | 2 - 3 | 457 | 69 | ||
FQ216583.1 | - | 2 - 3 | 417 | 69 | ||
FQ218392.1 | - | 2 - 3 | 400 | 69 | ||
FQ218618.1 | - | 2 - 3 | 463 | 69 | ||
FQ232448.1 | - | 2 - 3 | 417 | 69 | ||
FQ233915.1 | - | 2 - 3 | 324 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
