Assay Details
Gene Symbol: Fam96a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008327.1 | NP_001008328.1 | 3 - 4 | 476 | 62 | |
GenBank mRNA | BC086524.1 | - | 3 - 4 | 476 | 7303322 | 62 |
FQ210328.1 | - | 3 - 4 | 457 | 62 | ||
FQ215407.1 | - | 3 - 4 | 513 | 62 | ||
FQ216067.1 | - | 3 - 4 | 520 | 62 | ||
FQ216458.1 | - | 3 - 4 | 514 | 62 | ||
FQ216583.1 | - | 3 - 4 | 474 | 62 | ||
FQ218392.1 | - | 3 - 4 | 457 | 62 | ||
FQ218618.1 | - | 3 - 4 | 520 | 62 | ||
FQ225271.1 | - | 3 - 4 | 373 | 62 | ||
FQ232448.1 | - | 3 - 4 | 474 | 62 | ||
FQ233915.1 | - | 3 - 4 | 381 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
