Assay Details
Gene Symbol: Fam96a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008327.1 | NP_001008328.1 | 4 - 5 | 522 | 97 | |
GenBank mRNA | BC086524.1 | - | 4 - 5 | 522 | 7303322 | 97 |
FQ215407.1 | - | 4 - 5 | 559 | 97 | ||
FQ216458.1 | - | 4 - 5 | 560 | 97 | ||
FQ218392.1 | - | 4 - 5 | 503 | 97 | ||
FQ218618.1 | - | 4 - 5 | 566 | 97 | ||
FQ220244.1 | - | 4 - 5 | 482 | 97 | ||
FQ225271.1 | - | 4 - 5 | 419 | 97 | ||
FQ232448.1 | - | 4 - 5 | 520 | 97 | ||
FQ233915.1 | - | 4 - 5 | 427 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
