Assay Details
Gene Symbol: Ostm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029925.1 | NP_001025096.1 | 826 | 68 | ||
XM_006256577.2 | XP_006256639.1 | 4 - 5 | 868 | 68 | ||
GenBank mRNA | BC097978.1 | - | 826 | 7442765 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
