Assay Details
Gene Symbol: RGD1306954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008288.1 | NP_001008289.1 | 3 - 4 | 483 | 82 | |
GenBank mRNA | BC085340.1 | - | 3 - 4 | 483 | 7302628 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
