Assay Details
| Gene Symbol: Chrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134880.1 | NP_001128352.1 | 1 - 2 | 161 | 72 | |
| GenBank mRNA | FQ209946.1 | - | 1 - 2 | 162 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chrac1
Genomic Map