Assay Details
| Gene Symbol: Mroh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103364.1 | NP_001096834.1 | 7 - 8 | 751 | 57 | |
| GenBank mRNA | BC093388.1 | - | 7 - 8 | 751 | 7389659 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|

Mroh1
Genomic Map