Assay Details
| Gene Symbol: Tmem194a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271249.1 | NP_001258178.1 | 1 - 2 | 281 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem194a| Gene Symbol: Tmem194a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271249.1 | NP_001258178.1 | 1 - 2 | 281 | 131 | |
Set Membership: |
|