Assay Details
| Gene Symbol: Ctdsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241502.2 | XP_006241564.1 | 5 - 6 | 458 | 74 | |
| XM_008765431.1 | XP_008763653.1 | 1 - 2 | 1115 | 74 | ||
More Information
Set Membership: |
|

Ctdsp2
Genomic Map