Assay Details
| Gene Symbol: Cbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012119.1 | NP_001012119.1 | 1 - 2 | 84 | 69 | |
| XM_006241994.2 | XP_006242056.1 | 1 - 2 | 197 | 69 | ||
| GenBank mRNA | BC087122.1 | - | 1 - 2 | 84 | 7108347 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Cbx6
Genomic Map