Assay Details
| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011993.1 | NP_001011993.1 | 1 - 2 | 154 | 56 | |
| GenBank mRNA | BC087166.1 | - | 1 - 2 | 154 | 7135468 | 56 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pycrl
Genomic Map