Assay Details
| Gene Symbol: Pycrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011993.1 | NP_001011993.1 | 2 - 3 | 217 | 69 | |
| GenBank mRNA | BC087166.1 | - | 2 - 3 | 217 | 7135468 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Pycrl
Genomic Map