Assay Details
| Gene Symbol: Ddt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024131.1 | NP_077045.1 | 2 - 3 | 364 | 218 | |
| GenBank mRNA | FQ209890.1 | - | 2 - 3 | 381 | 218 | |
| FQ209898.1 | - | 2 - 3 | 336 | 218 | ||
| FQ210467.1 | - | 2 - 3 | 337 | 218 | ||
| FQ210521.1 | - | 2 - 3 | 380 | 218 | ||
| FQ220991.1 | - | 2 - 3 | 374 | 218 | ||
| Z36980.1 | - | 2 - 3 | 364 | 218 | ||
More Information
Set Membership: |
|

Ddt
Genomic Map