Assay Details
Gene Symbol: RT1-DOb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008846.1 | NP_001008846.1 | 2 - 3 | 367 | 119 | |
GenBank mRNA | KC222915.1 | - | 2 - 3 | 367 | 119 | |
KC222916.1 | - | 2 - 3 | 367 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
