Assay Details
| Gene Symbol: Slc16a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772941.1 | XP_008771163.1 | 1 - 2 | 297 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008772941.1 transcripts.
Set Membership: |
|

Slc16a9
Genomic Map