Assay Details
Gene Symbol: Ddhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033066.1 | NP_001028238.1 | 5 - 6 | 1409 | 61 | |
XM_006251769.2 | XP_006251831.1 | 7 - 8 | 1600 | 61 | ||
XM_006251770.2 | XP_006251832.1 | 6 - 7 | 1579 | 61 | ||
XM_006251771.2 | XP_006251833.1 | 7 - 8 | 1600 | 61 | ||
XM_006251772.2 | XP_006251834.1 | 6 - 7 | 1498 | 61 | ||
XM_006251773.2 | XP_006251835.1 | 5 - 6 | 1477 | 61 | ||
XM_006251774.2 | XP_006251836.1 | 6 - 7 | 1497 | 61 | ||
GenBank mRNA | BC103649.1 | - | 5 - 6 | 1409 | 7314879 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
