Assay Details
Gene Symbol: Ston1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182954.2 | NP_891999.2 | 1 - 2 | 285 | 70 | |
XM_008764479.1 | XP_008762701.1 | 2 - 3 | 373 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_182954.2, XM_008764479.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
