Assay Details
| Gene Symbol: Mpv17l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098240.1 | NP_001091710.1 | 5 - 6 | 656 | 67 | |
| XM_008764516.1 | XP_008762738.1 | 5 - 6 | 5503 | 67 | ||
| GenBank mRNA | BC091193.1 | - | 5 - 6 | 656 | 7373705 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Mpv17l
Genomic Map