Assay Details
Gene Symbol: Nemf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170586.1 | NP_001164057.1 | 11 | 991 | 144 | |
GenBank mRNA | FM071452.1 | - | 7 | 507 | 144 | |
FQ205447.1 | - | 8 | 611 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
