Assay Details
Gene Symbol: Immp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729645.3 | XP_002729691.1 | 6 - 7 | 553 | 102 | |
XM_006240059.2 | XP_006240121.1 | 5 - 6 | 795 | 102 | ||
XM_006240060.2 | XP_006240122.1 | 644 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
