Assay Details
| Gene Symbol: Mfsd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729615.3 | XP_002729661.1 | 6 - 7 | 991 | 64 | |
| XM_006239869.2 | XP_006239931.1 | 6 - 7 | 1018 | 64 | ||
| XM_006239870.2 | XP_006239932.1 | 5 - 6 | 892 | 64 | ||
More Information
Set Membership: |
|

Mfsd2b
Genomic Map