Assay Details
| Gene Symbol: Srp54a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053871.1 | NP_446323.1 | 10 - 11 | 1131 | 163 | |
| XM_006240097.2 | XP_006240159.1 | 10 - 11 | 1143 | 163 | ||
| GenBank mRNA | BC079117.1 | - | 10 - 11 | 1131 | 7126297 | 163 |
| FQ211580.1 | - | 10 - 11 | 1142 | 163 | ||
| FQ225417.1 | - | 10 - 11 | 1146 | 163 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srp54a
Genomic Map