Assay Details
Gene Symbol: Adi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199097.1 | NP_954528.1 | 2 - 3 | 251 | 109 | |
GenBank mRNA | AY346335.1 | - | 2 - 3 | 251 | 109 | |
BC092562.1 | - | 2 - 3 | 240 | 7326245 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
