Assay Details
| Gene Symbol: Ppp2r5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106740.1 | NP_001100210.1 | 11 - 12 | 1724 | 68 | |
| XM_006240222.2 | XP_006240284.1 | 13 - 14 | 1888 | 68 | ||
| XM_006240223.2 | XP_006240285.1 | 13 - 14 | 1886 | 68 | ||
| XM_006240224.2 | XP_006240286.1 | 12 - 13 | 1626 | 68 | ||
| XM_006240225.2 | XP_006240287.1 | 12 - 13 | 1535 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P050897 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P881262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164859 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P050897 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P881262 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P050897 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P881262 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P881262 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P050897 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P501783 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29965 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72372 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74643 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10890640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890635 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890644 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890641 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890630 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890642 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10890629 | - | - |
| Rat Genome 230 2.0 Array | 1388965_at | - | - |
| Rat Genome 230 2.0 Array | 1395826_at | - | - |
| Rat Genome 230 2.0 Array | 1379554_at | - | - |
| Rat Genome 230 2.0 Array | 1378530_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378530_at | - | - |
| Rat Genome 230 2.0 Array | 1379554_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392558_at | - | - |
| Rat Genome 230 2.0 Array | 1388965_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392558_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395826_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378530_at | - | - |
| Affymetrix HT Rat Focus Array | 1379554_at | - | - |
| Affymetrix HT Rat Focus Array | 1388965_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379554_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392558_at | - | - |
| Affymetrix HT Rat Focus Array | 1378530_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388965_at | - | - |
| Affymetrix HT Rat Focus Array | 1395826_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392558_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395826_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600019030.rn.1 | TC0600002681.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600019033.rn.1 | TC0600002681.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600019034.rn.1 | TC0600002681.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600019027.rn.1 | TC0600002681.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600019028.rn.1 | TC0600002681.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600019035.rn.1 | TC0600002681.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600019031.rn.1 | TC0600002681.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388965_PM_at | - | - |
| Rat Expression Set 230 | 1388965_at | - | - |
| Rat Expression Set 230 | 1378530_at | - | - |
| Rat Expression Set 230 | 1392558_PM_at | - | - |
| Rat Expression Set 230 | 1379554_at | - | - |
| Rat Expression Set 230 | 1392558_at | - | - |
| Rat Expression Set 230 | 1395826_at | - | - |
| Rat Expression Set 230 | 1378530_PM_at | - | - |
| Rat Expression Set 230 | 1379554_PM_at | - | - |
| Rat Expression Set 230 | 1395826_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10890629 | - | - |
| Rat Genome U34 Set | rc_AI101600_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21520751 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360812 | - | - |

Ppp2r5e
Genomic Map