Assay Details
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031655.1 | NP_113843.1 | 2 - 3 | 364 | 95 | |
GenBank mRNA | BC081763.1 | - | 2 - 3 | 350 | 7125782 | 95 |
FQ220594.1 | - | 2 - 3 | 379 | 95 | ||
FQ229501.1 | - | 2 - 3 | 356 | 95 | ||
U40260.1 | - | 2 - 3 | 331 | 95 | ||
X76985.1 | - | 2 - 3 | 364 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
