Assay Details
Gene Symbol: Sun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251471.2 | XP_006251533.1 | 7 - 8 | 870 | 68 | |
XM_008770406.1 | XP_008768628.1 | 7 - 8 | 4452 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
