Assay Details
| Gene Symbol: Akap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114606.1 | NP_001108078.1 | 12 - 13 | 1992 | 66 | |
| XM_006246509.2 | XP_006246571.1 | 12 - 13 | 3589 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akap10
Genomic Map