Assay Details
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107461.1 | NP_001100931.1 | 3 - 4 | 653 | 107 | |
XM_006227927.2 | XP_006227989.1 | 5 - 6 | 761 | 107 | ||
XM_006227928.2 | XP_006227990.1 | 5 - 6 | 761 | 107 | ||
XM_006227930.2 | XP_006227992.1 | 5 - 6 | 761 | 107 | ||
XM_008758757.1 | XP_008756979.1 | 3 - 4 | 655 | 107 | ||
XM_008758758.1 | XP_008756980.1 | 3 - 4 | 655 | 107 | ||
XM_008758759.1 | XP_008756981.1 | 4 - 5 | 698 | 107 | ||
GenBank mRNA | BC166440.1 | - | 5 - 6 | 692 | 7455355 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
