Assay Details
Gene Symbol: Tcp10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107461.1 | NP_001100931.1 | 6 - 7 | 990 | 92 | |
XM_006227928.2 | XP_006227990.1 | 9 - 10 | 1197 | 92 | ||
XM_008758757.1 | XP_008756979.1 | 7 - 8 | 1091 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
