Assay Details
| Gene Symbol: Rsph10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013867.1 | NP_001013889.1 | 7 - 8 | 936 | 52 | |
| XM_006248858.2 | XP_006248920.1 | 8 - 9 | 1763 | 52 | ||
| XM_006248859.2 | XP_006248921.1 | 8 - 9 | 1631 | 52 | ||
| XM_006248860.2 | XP_006248922.1 | 8 - 9 | 1763 | 52 | ||
| XM_006248861.2 | XP_006248923.1 | 7 - 8 | 1672 | 52 | ||
| GenBank mRNA | BC081935.1 | - | 7 - 8 | 936 | 7114952 | 52 |
| FQ212099.1 | - | 7 - 8 | 1043 | 52 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsph10b
Genomic Map