Assay Details
| Gene Symbol: Rsph10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013867.1 | NP_001013889.1 | 10 - 11 | 1406 | 63 | |
| XM_006248858.2 | XP_006248920.1 | 11 - 12 | 2233 | 63 | ||
| XM_006248859.2 | XP_006248921.1 | 11 - 12 | 2101 | 63 | ||
| XM_006248860.2 | XP_006248922.1 | 11 - 12 | 2233 | 63 | ||
| XM_006248861.2 | XP_006248923.1 | 10 - 11 | 2142 | 63 | ||
| GenBank mRNA | BC081935.1 | - | 10 - 11 | 1406 | 7114952 | 63 |
| FQ212099.1 | - | 10 - 11 | 1513 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsph10b
Genomic Map