Assay Details
| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034921.1 | NP_001030093.1 | 10 - 11 | 1263 | 76 | |
| GenBank mRNA | BC088466.1 | - | 420 | 76 | ||
| BC104691.1 | - | 10 - 11 | 1263 | 7107607 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P040386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727307 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50156 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040386 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P040386 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P040386 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P252906 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10101 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58260 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51785 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97422 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58929 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86525 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10708728 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708729 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708732 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708733 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708739 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708737 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708731 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708736 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708738 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708735 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708730 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10708727 | - | - |
| Rat Genome 230 2.0 Array | 1381448_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381448_at | - | - |
| Rat Genome 230 2.0 Array | 1392533_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392533_at | - | - |
| Affymetrix HT Rat Focus Array | 1392533_at | - | - |
| Affymetrix HT Rat Focus Array | 1392533_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381448_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381448_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100015866.rn.1 | TC0100002068.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100015863.rn.1 | TC0100002068.rn.1 | RTA TC |
| Rat Expression Set 230 | 1381448_PM_at | - | - |
| Rat Expression Set 230 | 1381448_at | - | - |
| Rat Expression Set 230 | 1392533_PM_at | - | - |
| Rat Expression Set 230 | 1392533_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708727 | - | - |
| Rat Genome U34 Set | rc_AI170861_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21817886 | - | - |
| ABI Rat Genome Survey Microarray | 22411746 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365786 | - | - |

Nars2
Genomic Map