Assay Details
| Gene Symbol: Tubb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109119.1 | NP_001102589.1 | 129 | 116 | ||
| XM_008771582.1 | XP_008769804.1 | 154 | 116 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109119.1, XM_008771582.1 transcripts.
Set Membership: |
|

Tubb2a
Genomic Map