Assay Details
| Gene Symbol: Ifitm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106314.1 | NP_001099784.1 | 2 - 3 | 350 | 161 | |
| XM_006230530.1 | XP_006230592.1 | 2 - 3 | 433 | 161 | ||
| GenBank mRNA | FQ221702.1 | - | 1 - 2 | 203 | 161 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifitm1
Genomic Map