Assay Details
| Gene Symbol: Ube2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037292.1 | NP_001032369.1 | 2 - 3 | 190 | 117 | |
| GenBank mRNA | U13176.1 | - | 2 - 3 | 190 | 117 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2d2
Genomic Map