Assay Details
| Gene Symbol: Hsd3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007719.3 | NP_001007720.3 | 4 - 5 | 465 | 104 | |
| GenBank mRNA | BC086578.1 | - | 4 - 5 | 465 | 7306702 | 104 |
| M38178.1 | - | 3 - 5 | 484 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd3b1
Genomic Map