Assay Details
Gene Symbol: Pgrmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF163321.1 | - | 1 - 2 | 387 | 145 | |
AJ005837.1 | - | 1 - 2 | 401 | 145 | ||
BC062073.1 | - | 1 - 2 | 390 | 5599449 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
