Assay Details
| Gene Symbol: LOC100910138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751350.3 | XP_003751398.1 | 2 - 3 | 208 | 156 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003751350.3 transcripts.
Set Membership: |
|

LOC100910138
Genomic Map