Assay Details
| Gene Symbol: RGD1303003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004225.1 | NP_001004225.1 | 7 | 139 | ||
| GenBank mRNA | BC079380.1 | - | 7 | 7110981 | 139 | |
| FQ213331.1 | - | 7 | 139 | |||
| FQ217777.1 | - | 7 | 139 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RGD1303003
Genomic Map