Assay Details
| Gene Symbol: Olr397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001000380.1 | NP_001000380.1 | 1 | 467 | 113 | |
| XM_006234064.2 | XP_006234126.1 | 2 | 804 | 113 | ||
| XM_006234065.2 | XP_006234127.1 | 2 | 801 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olr397
Genomic Map