Assay Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017171.1 | NP_058867.1 | 10 - 11 | 1780 | 106 | |
GenBank mRNA | AY642593.1 | - | 10 - 11 | 1594 | 106 | |
M15523.1 | - | 3 - 4 | 399 | 106 | ||
M18331.1 | - | 10 - 11 | 1780 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
