Assay Details
| Gene Symbol: Uba52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031687.3 | NP_113875.1 | 1 - 2 | 39 | 68 | |
| GenBank mRNA | BC061544.1 | - | 1 - 2 | 35 | 6922034 | 68 |
| BP480615.1 | - | 3 - 4 | 68 | |||
| FQ209944.1 | - | 1 - 2 | 31 | 68 | ||
| FQ212001.1 | - | 1 - 2 | 33 | 68 | ||
| FQ212205.1 | - | 1 - 2 | 31 | 68 | ||
| FQ217153.1 | - | 1 - 2 | 40 | 68 | ||
| FQ221631.1 | - | 1 - 2 | 31 | 68 | ||
| FQ222440.1 | - | 1 - 2 | 40 | 68 | ||
| FQ223258.1 | - | 1 - 2 | 31 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061544.1,NM_031687.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uba52
Genomic Map