Assay Details
| Gene Symbol: Prss30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199371.1 | NP_955403.1 | 1 - 2 | 94 | 83 | |
| XM_006245922.2 | XP_006245984.1 | 2 - 3 | 285 | 83 | ||
| XM_006245923.2 | XP_006245985.1 | 2 - 3 | 293 | 83 | ||
| XM_008767553.1 | XP_008765775.1 | 2 - 3 | 313 | 83 | ||
| GenBank mRNA | AB073023.1 | - | 1 - 2 | 94 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss30
Genomic Map